ContactUsLinkComponent
Test strips, cyanuric acid, swimming pool test
Test strips, cyanuric acid, swimming pool test
  MANA90710
 :  
undefined
Test strips, cyanuric acid, swimming pool test
  MANA90710
 :  
  • Pk:
    1
  • Description:
    Cyanuric acid test strips
  • Seq:
    03
  • Tests per kit:
    25
  • Range:
    0 - 300 mg/l
These test strips allow the quick and easy determination of the water quality in swimming pools. The test provides valuable results about the Cyanuric acid content.
  • Reliable result is obtained within 30 seconds.
  • Just dip, wait and read against multi-stage color scale
  • Always ready to use
  • No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
The test strip is dipped into the pool and moved back and forth 5 times.
Monitoring of the water quality allows adjusting the dosage of water conditioners in good time. The pool remains nice and clean.