Orders LinkContactUsLinkComponent
QUANTOFIX® Test Strips for Semi-quantitative Analysis of Low Water Hardness, AQUADUR® Sensitive
QUANTOFIX® Test Strips for Semi-quantitative Analysis of Low Water Hardness, AQUADUR® Sensitive
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Test Strips for Semi-quantitative Analysis of Low Water Hardness, AQUADUR® Sensitive
Supplier:  MACHEREY-NAGEL
Certificates
QUANTOFIX® Test Strips for Semi-quantitative Analysis of Low Water Hardness, AQUADUR® Sensitive
QUANTOFIX® Test Strips for Semi-quantitative Analysis of Low Water Hardness, AQUADUR® Sensitive
3 Options Available Below
Product Details & Documents
AQUADUR® sensitive test strips for semi-quantitative analysis of low water hardness, after amongst other things the softening of water.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are ideal to test low levels of water hardness. The easy 'dip and read' procedure provides reliable results within seconds.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • MANA932009
  • MANA932004
  • MANA932001
  • Method:
  • Total hardness - white → pink
  • Chlorine - yellow → violet
  • Peracetic acid - white → dark blue
  • Parameters:
  • All reagents included
  • All reagents included
  • All reagents included
  • Compliance:
  • No hazardous material
  • No hazardous material
  • No hazardous material
  • Contents:
  • 50 test strips in a tube, instruction leaflet
  • 100 test strips in a tube, instruction leaflet
  • 100 test strips in a tube, instruction leaflet
  • Instrument:
  • No
  • No
  • No
  • Platform:
  • Semi-quantitative test strips
  • Semi-quantitative test strips
  • Semi-quantitative test strips
  • Shelf life (months):
  • 30 (from production)
  • 30 (from production)
  • 30 (from production)
  • Weight:
  • 36 g
  • 50 g
  • 43 g
Specifications
Product Family Options
Product Information
  • Range
    Dimensions
    Tests per kit
    Pk
    Availability
    Price
  • MANA932009
    Diaquant® water hardness
    Diaquant® water hardness
    0 • 5 • 10 • 20 - ppm - CaCO₃
    38×38×120 mm (packaging)
    50 Tests
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 25,90 €
    25,90 €
    1 * 50 ST
       25,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA932009
    Method
    Total hardness - white → pink
    Parameters
    All reagents included
    Compliance
    No hazardous material
    Contents
    50 test strips in a tube, instruction leaflet
    Instrument
    No
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    36 g
  • MANA932004
    Diaquant® chlorine
    Diaquant® chlorine
    0 • 0,1 • 0,5 • 1 • 3 • 10 - mg/L - Cl₂
    43×126×40 mm (packaging)
    100 Tests
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 33,10 €
    33,10 €
    1 * 100 ST
       33,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA932004
    Method
    Chlorine - yellow → violet
    Parameters
    All reagents included
    Compliance
    No hazardous material
    Contents
    100 test strips in a tube, instruction leaflet
    Instrument
    No
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    50 g
  • MANA932001
    Diaquant® peracetic acid
    Diaquant® peracetic acid
    0 • 5 • 10 • 20 • 30 • 50 - mg/L - Peracetic acid
    40×40×120 mm (packaging)
    100 Tests
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 41,20 €
    41,20 €
    1 * 100 ST
       41,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA932001
    Method
    Peracetic acid - white → dark blue
    Parameters
    All reagents included
    Compliance
    No hazardous material
    Contents
    100 test strips in a tube, instruction leaflet
    Instrument
    No
    Platform
    Semi-quantitative test strips
    Shelf life (months)
    30 (from production)
    Weight
    43 g