ContactUsLinkComponent
Quantus® Protect, Cleanroom Coats with Zip Closure and Pockets
Quantus® Protect, Cleanroom Coats with Zip Closure and Pockets
Catalog # QNTS081-3PTWH0XLID
Supplier:  VWR Collection
Quantus® Protect, Cleanroom Coats with Zip Closure and Pockets
Catalog # QNTS081-3PTWH0XLID
Supplier:  VWR Collection
Specifications
  • Clothing Size:
    XL
  • Closure type:
    Zip closure
  • Garment application:
    Cleanroom
  • Gender:
    Unisex
  • Risk area:
    Anti-static
  • Colour:
    White
  • Seq:
    05
  • Fabric:
    Polyester/carbon
  • Sleeve or leg length:
    Long
  • Pk:
    1
Specifications
About this item
These coats have both zipper and pockets.
  • Hanger in collar
  • Stitched collar patch with labe
  • Antistatic knitted cuffs
  • One breast pocket and two hip pockets
  • Two way zip in front
The coats contain press studs on left breast for PES clipband and also lap seams for tight and secure seam-enclosure.