Order Entry
Austria
ContactUsLinkComponent
Multi-element standard in 75 blank oil: S-21:100 ppm, CONOSTAN®
Multi-element standard in 75 blank oil: S-21:100 ppm, CONOSTAN®
Catalog # PROOCON150-021-003
CAS Number:  
Multi-element standard in 75 blank oil: S-21:100 ppm, CONOSTAN®
Catalog # PROOCON150-021-003
CAS Number:  

Specifications

  • Concentration:
    100 µg/g
  • Matrix:
    Base Oil 75
  • Pk:
    100 g
  • Description:
    Multi-element standard (21 components) in oil

Specifications

About this item