MEM vitamin solution, 100X, Corning®
Artikelnr. 25-020-CI
Lieferant: Corning
CAS-Nummer:
Spezifikationen
- Pk:6
- Source:
- Species:
- Description:MEM Vitamin Solution, 100X
- Environmentally Preferable:
Spezifikationen
Über diesen Artikel
The addition of MEM vitamins to medium can improve growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells.
This concentrated supplement allows for efficient addition to media with no risk of dilution of media components.