Immunotag™ rat HbA1C (glycosylated hemoglobin/hemoglobin A1c) ELISA kit
Catalog # GENOIT7389
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rat
- Description:Immunotag™ rat HbA1C (glycosylated hemoglobin/hemoglobin A1c) ELISA kit, 1×96-well plate
- Detection range:1,563 - 100 ng/ml
- Sensitivity:0,938 ng/ml