Order Entry
Austria
ContactUsLinkComponent
Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Catalog # 95164-250ML
CAS Number:  
Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Catalog # 95164-250ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Description:
    Cesium chloride lanthanum chloride buffer

Specifications

About this item

TraceSELECT™, for flame atomic absorption spectrometry.

  • Packaged in high-quality containers appropriate for each product
  • The reagents are produced and bottled under clean-room conditionsions

Lanthanum chloride (LaCl3) 99,0 - 101,0 g/l
Cesium chloride (Cscl) 99,0 - 101,0 g/l
Density (D 20/4) 1,152 - 1,176
Calcium (Ca) max. 0,5 ppm