Order Entry
Austria
ContactUsLinkComponent
Davidson solution
Davidson solution
Catalog # APPCA3200.1000
Supplier:  PanReac AppliChem
CAS Number:  
Davidson solution
Catalog # APPCA3200.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Davidson solution

Specifications

About this item

Davidson solution is a rapid fixative.

Composition:
Deionised water: 347 ml/l
Acetic acid 100 %: 111 ml/l
Ethanol 99 %: 320 ml/l
Formaldehyde - Solution 10 % phosphate buffered: 222 ml/l