Oxidation reagent 0.02 M I₂ in 78/20/2 THF/pyridine/water (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR663-4X2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 L
- Seq:5114
- Environmentally Preferable:
- Pk Info:4x 2.5 L
Specifications
About this item
:
:253.81 g/mol
Boiling Pt:66 °C
Melting Pt:-108.5 °C
Melting Pt:–108.5 °C
Density:0.888 g/cm3
Flash Pt:-15 °C
Flash Pt:–15 °C
UN:1993
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355