Orders LinkContactUsLinkComponent
Erlenmeyer flasks, polymethylpentene
Erlenmeyer flasks, polymethylpentene
250 ml
Catalog # HACH2089846
Supplier:   Hach
Erlenmeyer flasks, polymethylpentene
Catalog # HACH2089846
Supplier:  Hach
Erlenmeyer flasks, polymethylpentene
Erlenmeyer flasks, polymethylpentene
250 ml
Price
36,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Ideal for titrations, preparation and storage of media.
  • Chemical resistance Strength
  • Autoclavable
Polymethylpentene flask with polypropylene screwcap features clarity (Do not engage threads during cycle).
Specifications
  • Capacity:
    250 ml
  • Seq:
    02