Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes, Rectangular

Storage Boxes, Rectangular

Supplier:   Gilac PROFESSIONNEL
Storage Boxes, Rectangular
Certificates
Storage Boxes, Rectangular

9 Options Available Below

Product Details & Documents

These rectangular storage boxes made from HDPE are ideal for sub-zero and ambient temperature storage and transporting raw materials.

  • No gaps or overlaps, rounded corners for easy cleaning
  • Machine washable
  • Optional lid for improved hygiene
  • Stackable with and without lid
  • Vertical lateral reinforcements for increased load support
  • Compatible with Gilac 4- and 6-wheel dollies

Ordering information: Lids need to be ordered separately.

Product Family Options

Product Information

  • Capacity
    Colour
    W×D×H
    Material
    Weight
    Availability
    Price
  • 15 l
    White
    400×300×215 mm
    HDPE
    1,09 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,50 €
    30,50 €
    1 * 1 ST
       30,50 €

  • 15 l
    Grey
    400×300×215 mm
    HDPE
    1,09 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,50 €
    30,50 €
    1 * 1 ST
       30,50 €

  • 35 l
    White
    600×400×217 mm
    HDPE
    1,6 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,70 €
    50,70 €
    1 * 1 ST
       50,70 €

  • 35 l
    Grey
    600×400×217 mm
    HDPE
    1,6 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,70 €
    50,70 €
    1 * 1 ST
       50,70 €

  • 55 l
    White
    600×400×315 mm
    HDPE
    2,2 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 63,50 €
    63,50 €
    1 * 1 ST
       63,50 €

  • 55 l
    Grey
    600×400×315 mm
    HDPE
    2,2 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,60 €
    65,60 €
    1 * 1 ST
       65,60 €

Lids

Product Information

  • Capacity
    Colour
    W×D×H
    Material
    Weight
    Availability
    Price
  • CORAG118921
    Lid for 12 and 15 l containers
    Lid for 12 and 15 l containers
    White
    400×300×30 mm
    HDPE
    0,32 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €

  • CORAG118977
    Lid for 12 and 15 l containers
    Lid for 12 and 15 l containers
    Grey
    400×300×30 mm
    HDPE
    0,32 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €

  • CORAG193377
    Lid for 25, 35 and 55 l containers
    Lid for 25, 35 and 55 l containers
    Grey
    600×400×35 mm
    HDPE
    0,86 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.