Order Entry
Austria
ContactUsLinkComponent
 
Laboratory grinding mill, MultiDrive control
 :  IKA
 
undefined
Laboratory grinding mill, MultiDrive control
 :  IKA

 

  • Catalog No:
  • 431-0295
  • 431-0298
  • 412-0325
  • Model:
  • MultiDrive control
  • Max. size of feed granules:
  • 7 mm
  • Duty cycle on/off:
  • 5/10 min
  • Electrical:
  • 220 - 240/100 - 120 V, 50/60 Hz
  • Grinding chamber material:
  • Stainless steel
  • Motor rating input/output:
  • 1000/800 W
  • Speed range:
  • 3000 - 20000 min⁻¹
  • Usable volume:
  • 2000 ml
  • W×D×H:
  • 300×250×450 mm
  • Weight:
  • 10 kg

 

Frequently Bought Together