Order Entry
Austria
ContactUsLinkComponent
Ethidium homodimer I 2 mM in DMSO used in nuclear staining
Ethidium homodimer I 2 mM in DMSO used in nuclear staining
Catalog # 40014.
Supplier:  Biotium
CAS Number:  
undefined
Ethidium homodimer I 2 mM in DMSO used in nuclear staining
Catalog # 40014.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    0,5 mL
  • Seq:
    2040

Specifications

About this item

 :
 :856.75 g/mol
Storage Temperature:Refrigerator
CAS Number:61926-22-5
Formula:C₄₆H₅₀Cl₄N₈
MDL Number:MFCD00210948