Indo-1 pentapotassium salt ≥90% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50041.
Supplier: Biotium
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:840.05 g/mol
Storage Temperature:Refrigerator
CAS Number:132319-56-3
Formula:C₃₂H₂₆K₅N₃O₁₂