BAPTA-AM ≥98% (by TLC) ion chelator, for calcium determination
Supplier: Biotium
Certificates
About this item
CAS Number:126150-97-8
:
Formula:C₃₄H₄₀N₂O₁₈
MDL Number:MFCD00036696
:764.69 g/mol
Storage Temperature:Freezer
Frequently Bought Together