0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Hardness/TSS - Whole Volume, (CRM), Supelco®
Hardness/TSS - Whole Volume, (CRM), Supelco®
Catalog # QC3074-500ML
Supplier:
Merck
CAS Number:
Expand image
Hardness/TSS - Whole Volume, (CRM), Supelco®
Catalog # QC3074-500ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
500 mL
Description:
Hardness/TSS - Whole Volume
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details