Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Dissolved Oxygen - Whole Volume, (CRM), Supelco®
Dissolved Oxygen - Whole Volume, (CRM), Supelco®
Dissolved Oxygen - Whole Volume, (CRM), Supelco®
Catalog # QC3077-500ML
Supplier:
Merck
CAS Number:
Certificates
Expand image
Dissolved Oxygen - Whole Volume, (CRM), Supelco®
Catalog # QC3077-500ML
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
500 mL
Description:
Dissolved Oxygen - Whole Volume
Type:
Certified reference material
Specifications
About this item
Product Details
Documentation
Product Details
Documentation