Order Entry
Austria
ContactUsLinkComponent
Hardness calibration standard, calcium 1000 mg/L, (CRM), Supelco®
Hardness calibration standard, calcium 1000 mg/L, (CRM), Supelco®
Catalog # HARD1000-500ML
Supplier:  Merck
CAS Number:  
Hardness calibration standard, calcium 1000 mg/L, (CRM), Supelco®
Catalog # HARD1000-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Calcium 1000 mg/l
  • Pk:
    500 mL
  • Description:
    Hardness calibration standard
  • Type:
    Certified reference material

Specifications

About this item