Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Trace metals - sewage amended soil, (CRM), Supelco®
Trace metals - sewage amended soil, (CRM), Supelco®
Trace metals - sewage amended soil, (CRM), Supelco®
Catalog # CRM005-50G
Supplier:
Merck
CAS Number:
Expand image
Trace metals - sewage amended soil, (CRM), Supelco®
Catalog # CRM005-50G
Supplier:
Merck
CAS Number:
Specifications
Pk:
50 g
Description:
Trace Metals - Sewage Amended Soil
Pk Info:
Pkg of 50 g
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details