EPA 8260 Internal standards mix, (CRM), Supelco®
Catalog # CRM48958
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 µg/ml each component in methanol
- Pk:1 mL
- Description:EPA 8260 Internal Standards Mix
- Type:Certified reference material
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.