0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Anions - Sandy loam 1, (CRM), Supelco®
Anions - Sandy loam 1, (CRM), Supelco®
Anions - Sandy loam 1, (CRM), Supelco®
Catalog # CRM700-50G
Supplier:
Merck
CAS Number:
Expand image
Anions - Sandy loam 1, (CRM), Supelco®
Catalog # CRM700-50G
Supplier:
Merck
CAS Number:
Specifications
Pk:
50 g
Description:
Anions - Sandy Loam 1
Pk Info:
Pkg of 50 g
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details