Order Entry
Austria
ContactUsLinkComponent
Petri dishes, glass, with grid
Petri dishes, glass, with grid
Catalog # HECH41044010
undefined
Petri dishes, glass, with grid
Catalog # HECH41044010

Specifications

  • Ø:
    100 mm
  • Description:
    Petri dishes with 10×10 mm grid
  • Height:
    20 mm
  • Pk:
    1

Specifications

About this item

Petri dishes made of glass with grid and cover.