Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PMA (Propidium monoazide)
PMA (Propidium monoazide) DNA/RNA binding dye
PMA (Propidium monoazide) DNA/RNA binding dye
PMA (Propidium monoazide) DNA/RNA binding dye
Catalog # 40013.
Supplier:
Biotium
CAS Number:
Certificates
Expand image
PMA (Propidium monoazide) DNA/RNA binding dye
Catalog # 40013.
Supplier:
Biotium
CAS Number:
Certificates
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
Documentation
:
:
440.582 g/mol
Storage Temperature:
Freezer
Formula:
C₂₇H₃₂N₆²⁺
Product Details
Documentation