Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Succinimidyl ester methylene blue
Succinimidyl ester methylene blue DNA/RNA binding dye
Succinimidyl ester methylene blue DNA/RNA binding dye
Succinimidyl ester methylene blue DNA/RNA binding dye
Catalog # 40075.
Supplier:
Biotium
CAS Number:
Expand image
Succinimidyl ester methylene blue DNA/RNA binding dye
Catalog # 40075.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
533 g/mol
Storage Temperature:
Freezer
Formula:
C₂₄H₂₆BrN₄O₄S
Product Details