Order Entry
Austria
ContactUsLinkComponent
 
High Temperature Viscosity Standards, Supelco®
Catalog # SIALN100HT-500ML
Supplier:  Merck
CAS Number:  
High Temperature Viscosity Standards, Supelco®
Catalog # SIALN100HT-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    N100HT
  • Viscosity:
    100 °C: 11,77 mm²/s
    150 °C: 4,486 mm²/s

Specifications

About this item

  • Temperature range: 100 to 150 °C
  • Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/ml) given at all temperatures
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
Certifications: Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation.