Order Entry
Austria
ContactUsLinkComponent
Organic reference standards, (+/-)-Geosmin solution, (CRM), TraceCERT®, Supelco®
Organic reference standards, (+/-)-Geosmin solution, (CRM), TraceCERT®, Supelco®
Catalog # CRM47522
Supplier:  Merck
CAS Number:  
Organic reference standards, (+/-)-Geosmin solution, (CRM), TraceCERT®, Supelco®
Catalog # CRM47522
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    16423-19-1
  • Description:
    (+/-)-Geosmin solution
  • Environmentally Preferable:
  • Description:
    (+/-)-Geosmin solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item