Eosin-5-isothiocyanate amine-reactive fluorescent dye
Catalog # 90091.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
:704.96 g/mol
Storage Temperature:Freezer
CAS Number:60520-47-0
Formula:C₂₁H₇Br₄NO₅S
MDL Number:MFCD00082502