Order Entry
Austria
ContactUsLinkComponent
6-Carboxyrhodamine 110 succinimidyl ester ≥90% (by HPLC) amine-reactive fluorescent dye
6-Carboxyrhodamine 110 succinimidyl ester ≥90% (by HPLC) amine-reactive fluorescent dye
Catalog # 90031.
Supplier:  Biotium
CAS Number:  
undefined
6-Carboxyrhodamine 110 succinimidyl ester ≥90% (by HPLC) amine-reactive fluorescent dye
Catalog # 90031.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :600.67 g/mol
Storage Temperature:Freezer
Formula:C₃₃H₃₆N₄O₇