Order Entry
Austria
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥98% (by TLC) amine-reactive fluorescent dye
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90020.
Supplier:  Biotium
CAS Number:  
undefined
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90020.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :389.381 g/mol
Storage Temperature:Freezer
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063