Order Entry
Austria
ContactUsLinkComponent
 
5-Carboxyrhodamine 110-X succinimidyl ester amine-reactive fluorescent dye
Catalog # 90014.
Supplier:  Biotium
CAS Number:  
undefined
5-Carboxyrhodamine 110-X succinimidyl ester amine-reactive fluorescent dye
Catalog # 90014.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :584.58 g/mol
Storage Temperature:Freezer
Formula:C₃₁H₂₈N₄O₈