Supelco® TraceCERT® Organic Reference Standard, ʟ-Tyrosine
Catalog # 91515-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:60-18-4
- Description:ʟ-Tyrosine
- Storage conditions:15...25 °C
- Shipping conditions:STD
- MDL No.:MFCD00002606
- M-Clarity quality level:MQ300
- Synonym(s):(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid; 3-(4-Hydroxyphenyl)-ʟ-alanine
- Melting point:>300 °C (dec.) (lit.)
- Formula:4-(HO)C₆H₄CH₂CH(NH₂)CO₂H
- Molecular weight:181,19
Specifications
About this item
- GHS signal: GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.