Order Entry
Austria
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, ʟ-Ascorbic acid
  57803-100MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, ʟ-Ascorbic acid
  57803-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    50-81-7
  • Description:
    ʟ-Ascorbic acid
  • Storage conditions:
    20...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00064328
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    ʟ-Threoascorbic acid; Antiscorbutic factor; Vitamin C
  • Melting point:
    190 - 194 °C (dec.)
  • Molecular weight:
    176,12

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.