Order Entry
Austria
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Glycine
  76524-100MG
 :  Merck
 :  
undefined
Supelco® TraceCERT® Organic Reference Standard, Glycine
  76524-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    56-40-6
  • Description:
    Glycine
  • Storage conditions:
    20...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008131
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Aminoacetic acid; Aminoethanoic acid; Glycocoll
  • Melting point:
    240 °C (dec.) (lit.)
  • Formula:
    NH₂CH₂COOH
  • Molecular weight:
    75,07

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.