Supelco® Organic Reference Standard, (±)-Β-Hexabromocyclododecane
01986-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:134237-51-7
- Description:(±)-β-Hexabromocyclododecane
- Storage conditions:2...30 °C
- Hazard class description:H361fd - H362 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(1RS,2SR,5RS,6RS,9RS,10SR)-1,2,5,6,9,10-Hexabromocyclododecane; (±)-β-HBCD
- Melting point:152...158 °C
- Molecular weight:641,70
- Flash point:Not applicable
- GHS signal: GHS07
: P202 - P260 - P263 - P264 - P273 - P308 + P313