Order Entry
Austria
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, Tapentadol-D3 hydrochloride solution, (CRM)
  T-059-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, Tapentadol-D3 hydrochloride solution, (CRM)
  T-059-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    1435782-38-9
  • Description:
    Tapentadol-D3 hydrochloride solution
  • Description:
    Tapentadol-D3 hydrochloride solution
  • Pk Info:
    ampoule of 1 ml