Supelco® Drug and metabolite standards, N-desmethyl-cis-tramadol-D3 hydrochloride solution, (CRM)
Catalog # D-110-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1261398-09-7
- Description:N-Desmethyl-cis-tramadol-D3 hydrochloride solution
- Description:N-Desmethyl-cis-tramadol-D3 hydrochloride solution
- Pk Info:ampoule of 1 ml