Supelco® Drug and metabolite standards, naltrexone-D3 solution, (CRM)
N-047-1ML
: Merck
:
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1261080-26-5
- Description:Naltrexone-D3 solution
- Description:Naltrexone-D3 solution
- Pk Info:ampoule of 1 ml