Supelco® Drug and metabolite standards, nalbuphine-D3 solution, (CRM)
Catalog # N-056-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- Description:Nalbuphine-D3 solution
- Pk Info:ampoule of 1 ml