Order Entry
Austria
ContactUsLinkComponent
Supelco® Drug and metabolite standards, JWH-210 4-Hydroxypentyl metabolite-D5 solution, (CRM)
Supelco® Drug and metabolite standards, JWH-210 4-Hydroxypentyl metabolite-D5 solution, (CRM)
Catalog # S-057-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, JWH-210 4-Hydroxypentyl metabolite-D5 solution, (CRM)
Catalog # S-057-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    JWH-210 4-Hydroxypentyl metabolite-D5 solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item