Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Supelco® Drug and metabolite standards, Etizolam-D3 solution, (CRM)
Supelco® Drug and metabolite standards, Etizolam-D3 solution, (CRM)
Supelco® Drug and metabolite standards, Etizolam-D3 solution, (CRM)
E-082-1ML
:
Merck
:
Supelco® Drug and metabolite standards, Etizolam-D3 solution, (CRM)
E-082-1ML
:
Merck
:
Concentration:
100 µg/ml in methanol
Pk:
1 mL
Standards:
Certified reference material
Description:
Etizolam-D3 solution
Pk Info:
ampoule of 1 ml