Supelco® Drug and metabolite standards, Diphenhydramine-D3 solution, (CRM)
Catalog # D-017-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:170082-18-5
- Description:Diphenhydramine-D3 solution
- Description:Diphenhydramine-D3 solution
- Pk Info:ampoule of 1 ml