Order Entry
Austria
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, Dextrorphan-d3, (CRM)
  D-041-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, Dextrorphan-d3, (CRM)
  D-041-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    524713-57-3
  • Description:
    Dextrorphan-d3
  • Description:
    Dextrorphan-d3
  • Pk Info:
    ampoule of 1 ml