Supelco® Drug and metabolite standards, Dabigatran-D3 solution, (CRM)
Catalog # D-099-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml (Acetonitrile with 10% 0.01 N HCl)
- Pk:1 mL
- Standards:Certified reference material
- Description:Dabigatran-D3 solution
- Pk Info:ampoule of 1 ml