Drug and metabolite standards, 6beta-Naltrexol-D3 solution, (CRM), Supelco®
N-081-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1435727-11-9
- Description:6beta-Naltrexol-D3 solution
- Description:6beta-Naltrexol-D3 solution
- Pk Info:ampoule of 1 ml