Order Entry
Austria
ContactUsLinkComponent
Drug and metabolite standards, (1S,2S)-(+)-pseudoephedrine-d3 hydrochloride solution, (CRM), Supelco®
Drug and metabolite standards, (1S,2S)-(+)-pseudoephedrine-d3 hydrochloride solution, (CRM), Supelco®
Catalog # P-056-1ML
Supplier:  Merck
CAS Number:  
undefined
Drug and metabolite standards, (1S,2S)-(+)-pseudoephedrine-d3 hydrochloride solution, (CRM), Supelco®
Catalog # P-056-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    284665-25-4
  • Description:
    (1S,2S)-(+)-Pseudoephedrine-d3 hydrochloride solution
  • Description:
    (1S,2S)-(+)-Pseudoephedrine-d3 hydrochloride solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item