Drug and metabolite standards, (+/-)-Norpseudoephedrine-D3 hydrochloride solution, (CRM), Supelco®
N-087-1ML
: Merck
:
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- Description:(+/-)-Norpseudoephedrine-D3 hydrochloride solution
- Pk Info:ampoule of 1 ml