Drug and metabolite standards, (+/-)-methcathinone-D3 hydrochloride solution, (CRM), Supelco®
Catalog # M-189-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1246819-12-4
- Description:(+/-)-Methcathinone-D3 hydrochloride solution
- Description:(+/-)-Methcathinone-D3 hydrochloride solution