Order Entry
Austria
ContactUsLinkComponent
Food standards, Patulin solution, (CRM), TraceCERT®, Supelco®
Food standards, Patulin solution, (CRM), TraceCERT®, Supelco®
Catalog # CRM46914
Supplier:  Merck
CAS Number:  
undefined
Food standards, Patulin solution, (CRM), TraceCERT®, Supelco®
Catalog # CRM46914
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in chloroform
  • Pk:
    1 mL
  • CAS No.:
    149-29-1
  • Description:
    Patulin solution
  • Description:
    Patulin solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item

Patulin is a mycotoxin produced by several mold species. It has been found in fruit, especially apples, fruit juice, cider and stewed fruits. Because of its highly toxic properties, this compound has been the focus of many studies.

Caution: Storage temperature: Freezer −20 °C