Order Entry
Austria
ContactUsLinkComponent
Multi-element anionic standard, anions-whole volume, for ion chromatography, Supelco®
Multi-element anionic standard, anions-whole volume, for ion chromatography, Supelco®
Catalog # QC3060-500ML
Supplier:  Merck
CAS Number:  
Multi-element anionic standard, anions-whole volume, for ion chromatography, Supelco®
Catalog # QC3060-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pk:
    500 mL
  • Description:
    Anions - whole volume
  • Contents:
    10 analytes

Specifications

About this item

  • 10 analytes
Caution: Store at room temperature.