Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Sealing Rings
Solvent resistant O-ring
Solvent resistant O-ring
Solvent resistant O-ring
18-6300-01
:
Cytiva
Solvent resistant O-ring
18-6300-01
:
Cytiva
Description:
Solvent resistant O-ring, 11,3×2,4 mm, perfluoro elastomer
Pk:
3